Adamantane, 99+%, Thermo Scientific Chemicals
Adamantane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Adamantane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 146110050
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.20 
Save 2.55 (12%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.20 
Save 2.55 (12%)
Add to cart
Adamantane, 99+%, Thermo Scientific Chemicals
Catalog number146110050
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.20 
Save 2.55 (12%)
-
Add to cart
Chemical Identifiers
IUPAC Nameadamantane
Molecular FormulaC10H16
InChI KeyORILYTVJVMAKLC-UHFFFAOYSA-N
SMILESC1C2CC3CC1CC(C2)C3
Molecular Weight (g/mol)136.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
GC>=99.0 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only