Adamantane, 99+%
Adamantane, 99+%
Adamantane, 99+%
Thermo Scientific Chemicals

Adamantane, 99+%

CAS: 281-23-2 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 146110250
Price (USD)
41.65
Online Exclusive
45.80
Save 4.15 (9%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.80
Save 4.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS281-23-2
IUPAC Nameadamantane
Molecular FormulaC10H16
InChI KeyORILYTVJVMAKLC-UHFFFAOYSA-N
SMILESC1C2CC3CC1CC(C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
GC>=99.0 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link