N-Acetylethanolamine, 96%, Thermo Scientific Chemicals
N-Acetylethanolamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetylethanolamine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number 146201000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-26-7
IUPAC NameN-(2-hydroxyethyl)acetamide
Molecular FormulaC4H9NO2
InChI KeyPVCJKHHOXFKFRP-UHFFFAOYSA-N
SMILESCC(=O)NCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4720 to 1.4740 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only