3-Amino-1,2,4-triazine, 97%, Thermo Scientific Chemicals
3-Amino-1,2,4-triazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1,2,4-triazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 146230500
Price (USD)/ Each
312.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
312.00
Add to cart
3-Amino-1,2,4-triazine, 97%, Thermo Scientific Chemicals
Catalog number146230500
Price (USD)/ Each
312.00
-
Add to cart
Chemical Identifiers
CAS1120-99-6
IUPAC Name1,2,4-triazin-3-amine
Molecular FormulaC3H4N4
InChI KeyMJIWQHRXSLOUJN-UHFFFAOYSA-N
SMILESNC1=NC=CN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point173°C to 178°C
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Appearance (Color)Brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only