3-Benzyloxyaniline, 98%, Thermo Scientific Chemicals
3-Benzyloxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Benzyloxyaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 146260050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1484-26-0
IUPAC Name3-(benzyloxy)aniline
Molecular FormulaC13H13NO
InChI KeyIGPFOKFDBICQMC-UHFFFAOYSA-N
SMILESNC1=CC=CC(OCC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point62°C to 66°C
Appearance (Color)Beige to tan
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only