1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Thermo Scientific Chemicals

1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number 146310500
Price (USD)/ Each
631.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
743.00 
Save 111.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
631.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
743.00 
Save 111.35 (15%)
Add to cart
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Catalog number146310500
Price (USD)/ Each
631.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
743.00 
Save 111.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular FormulaC15H11N3O
InChI KeyRAXUMGMWXZYADR-SDXDJHTJSA-N
SMILESO=C1C=CC2=CC=CC=C2C1=NNC1=CC=CC=N1
Molecular Weight (g/mol)249.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red-brown to brown
Titration with HClO4>=97.5 %
Infrared spectrumConforms
Suitability(as complexom. indicator) Passes test
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only