Thermo Scientific Chemicals

Benzothiazole, 97%, Thermo Scientific Chemicals

Catalog number: 146390010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Benzothiazole, 97%, Thermo Scientific Chemicals

Catalog number: 146390010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 146390010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-16-9
IUPAC Name
1,3-benzothiazole
Molecular Formula
C7H5NS
InChI Key
IOJUPLGTWVMSFF-UHFFFAOYSA-N
SMILES
S1C=NC2=CC=CC=C12
Appearance (Color)
Clear yellow-brown to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.6410 to 1.6440 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text