Benzothiazole, 97%, Thermo Scientific Chemicals
Benzothiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzothiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 146390050
Price (USD)/ Each
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,3-benzothiazole
Molecular FormulaC7H5NS
InChI KeyIOJUPLGTWVMSFF-UHFFFAOYSA-N
SMILESS1C=NC2=CC=CC=C12
Molecular Weight (g/mol)135.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow-brown to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.6410 to 1.6440 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only