Thermo Scientific Chemicals

3-Chloro-2-methylaniline 99%, Thermo Scientific Chemicals

Catalog number: 146495000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Chloro-2-methylaniline 99%, Thermo Scientific Chemicals

Catalog number: 146495000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 146495000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-60-5
IUPAC Name
3-chloro-2-methylaniline
Molecular Formula
C7H8ClN
InChI Key
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
SMILES
CC1=C(N)C=CC=C1Cl
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5870 to 1.5890 (20°C, 589 nm)
Appearance (Color)
Clear yellow to red to red-brown
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text