Thermo Scientific Chemicals

Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Catalog number: 146570050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Catalog number: 146570050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146570050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3218-02-8
IUPAC Name
1-cyclohexylmethanamine
Molecular Formula
C7H15N
InChI Key
AVKNGPAMCBSNSO-UHFFFAOYSA-N
SMILES
NCC1CCCCC1
Appearance (Form)
Liquid
Refractive index
1.4610 to 1.4630 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text