Thermo Scientific Chemicals

2,4-Diaminotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 146600250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Diaminotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 146600250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146600250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-80-7
IUPAC Name
4-methylbenzene-1,3-diamine
Molecular Formula
C7H10N2
InChI Key
VOZKAJLKRJDJLL-UHFFFAOYSA-N
SMILES
CC1=CC=C(N)C=C1N
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or flakes and/or chunks
HPLC
>=97.5 %
Appearance (Color)
Dark grey to brown
Melting point
95°C to 102°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text