Thermo Scientific Chemicals

2,5-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 146690050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,5-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 146690050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 146690050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-78-3
IUPAC Name
2,5-dimethylaniline
Molecular Formula
C8H11N
InChI Key
VOWZNBNDMFLQGM-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)C(N)=C1
Appearance (Form)
Liquid
Refractive index
1.5582 to 1.5602 (20°C, 589 nm)
Appearance (Color)
Clear yellow to red-brown
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text