4-Fluorophenylacetonitrile, 99%, Thermo Scientific Chemicals
4-Fluorophenylacetonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorophenylacetonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 146720250
Price (USD)/ Each
53.40
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
53.40
Add to cart
4-Fluorophenylacetonitrile, 99%, Thermo Scientific Chemicals
Catalog number146720250
Price (USD)/ Each
53.40
-
Add to cart
Chemical Identifiers
CAS459-22-3
IUPAC Name2-(4-fluorophenyl)acetonitrile
Molecular FormulaC8H6FN
InChI KeyJHQBLYITVCBGTO-UHFFFAOYSA-N
SMILESFC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4990 to 1.5020 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only