Thermo Scientific Chemicals

1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals

Catalog number: 146730100
10 L, Metal Drum, Each
Thermo Scientific Chemicals

1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals

Catalog number: 146730100
10 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 146730100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-64-2
IUPAC Name
1,2,3,4-tetrahydronaphthalene
Molecular Formula
C10H12
InChI Key
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
SMILES
C1CCC2=CC=CC=C2C1
Infrared spectrum
Conforms
GC
>=98.0 %
Appearance (Color)
Clear colorless to light yellow
Appearance
May darken on storage
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text