Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
Packaging:
View all
Plastic Drum
Catalog number 146750250
Price (USD)/ Each
843.65
Online exclusive
937.00 
Save 93.35 (10%)
-
Add to cart
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
843.65
Online exclusive
937.00 
Save 93.35 (10%)
Add to cart
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number146750250
Price (USD)/ Each
843.65
Online exclusive
937.00 
Save 93.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
Molecular Weight (g/mol)62.07
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Acidity (CH3COOH)=<0.002 %
Appearance (Form)Clear viscous liquid
GC>=99.0 %
Water=<0.1 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only