Thermo Scientific Chemicals

N-Methylcyclohexylamine, 98%, Thermo Scientific Chemicals

Catalog number: 146992500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

N-Methylcyclohexylamine, 98%, Thermo Scientific Chemicals

Catalog number: 146992500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 146992500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-60-7
IUPAC Name
N-methylcyclohexanamine
Molecular Formula
C7H15N
InChI Key
XTUVJUMINZSXGF-UHFFFAOYSA-N
SMILES
CNC1CCCCC1
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow to brown
Infrared spectrum
Conforms
Refractive index
1.4535 to 1.4575 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text