Thermo Scientific Chemicals

Pentafluorobenzenesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 147110250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Pentafluorobenzenesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 147110250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 147110250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
832-53-1
IUPAC Name
pentafluorobenzene-1-sulfonyl chloride
Molecular Formula
C6ClF5O2S
InChI Key
UOJCTEGNHXRPKO-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C(F)=C1F)S(Cl)(=O)=O
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4780 to 1.4800 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text