Thermo Scientific Chemicals

Pentafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 147120250
25 g, Each
Thermo Scientific Chemicals

Pentafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 147120250
25 g, Each
Quantity
Catalog number: 147120250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2251-50-5
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.4520 to 1.4540 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text