Thermo Scientific Chemicals

Ethyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 147232500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 147232500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 147232500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-40-0
IUPAC Name
ethyl 4-methylbenzene-1-sulfonate
Molecular Formula
C9H12O3S
InChI Key
VRZVPALEJCLXPR-UHFFFAOYSA-N
SMILES
CCOS(=O)(=O)C1=CC=C(C)C=C1
GC
>=97.5 %
Refractive index
1.5100 to 1.5120 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light brown
Appearance (Form)
Liquid after melting
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text