Thermo Scientific Chemicals

n-Propyltrichlorosilane, 98%, Thermo Scientific Chemicals

Catalog number: 147410010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

n-Propyltrichlorosilane, 98%, Thermo Scientific Chemicals

Catalog number: 147410010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 147410010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-57-1
IUPAC Name
trichloro(propyl)silane
Molecular Formula
C3H7Cl3Si
InChI Key
DOEHJNBEOVLHGL-UHFFFAOYSA-N
SMILES
CCC[Si](Cl)(Cl)Cl
Infrared spectrum
Conforms
Refractive index
1.4290 to 1.4310 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text