Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals

Catalog number: 147490250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals

Catalog number: 147490250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 147490250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
111-18-2
IUPAC Name
[6-(dimethylamino)hexyl]dimethylamine
Molecular Formula
C10H24N2
InChI Key
TXXWBTOATXBWDR-UHFFFAOYSA-N
SMILES
CN(C)CCCCCCN(C)C
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4350 to 1.4370 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text