1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 147540250
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
Catalog number147540250
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS920-66-1
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.50 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only