1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure

CAS: 920-66-1 | C3H2F6O | 168.038 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 147541000
Price (EUR)
232,05
Online Exclusive
273,00
Save 40,95 (15%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
232,05
Online Exclusive
273,00
Save 40,95 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol)168.04
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.50 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link