1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 147580010
Price (USD)/ Each
36.50
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
36.50
Add to cart
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
Catalog number147580010
Price (USD)/ Each
36.50
-
Add to cart
Chemical Identifiers
IUPAC Name1,3-diphenyl-2-benzofuran
Molecular FormulaC20H14O
InChI KeyZKSVYBRJSMBDMV-UHFFFAOYSA-N
SMILESO1C(=C2C=CC=CC2=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)270.33
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point128°C to 137°C
Appearance (Color)Yellow to green
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only