1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 147580050
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
Add to cart
1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals
Catalog number147580050
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5471-63-6
IUPAC Name1,3-diphenyl-2-benzofuran
Molecular FormulaC20H14O
InChI KeyZKSVYBRJSMBDMV-UHFFFAOYSA-N
SMILESO1C(=C2C=CC=CC2=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point128°C to 137°C
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)Yellow to green
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only