Thermo Scientific Chemicals

Triphenyl phosphate, 99+%, Thermo Scientific Chemicals

Catalog number: 147670010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Triphenyl phosphate, 99+%, Thermo Scientific Chemicals

Catalog number: 147670010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 147670010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-86-6
IUPAC Name
triphenyl phosphate
Molecular Formula
C18H15O4P
InChI Key
XZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILES
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Flakes and/or pastilles
Water
=<0.1 %
Infrared spectrum
Conforms
Melting point
47°C to 53°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text