Triphenyl phosphate, 99+%, Thermo Scientific Chemicals
Triphenyl phosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triphenyl phosphate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 147670025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametriphenyl phosphate
Molecular FormulaC18H15O4P
InChI KeyXZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILESO=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
Molecular Weight (g/mol)326.29
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes and/or pastilles
Water=<0.1 %
Infrared spectrumConforms
Melting point47°C to 53°C
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only