1x1 image pixel for data collection
Cyclohexanol, 98%
Cyclohexanol, 98%
Cyclohexanol, 98%
Thermo Scientific Chemicals

Cyclohexanol, 98%

CAS: 108-93-0 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 147680025
Price (USD)
113.65
Special offer
Online exclusive
Ends: 26-Sep-2025
140.00
Save 26.35 (19%)
Each
In stock
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 26-Sep-2025
140.00
Save 26.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS108-93-0
IUPAC Namecyclohexanol
Molecular FormulaC6H12O
InChI KeyHPXRVTGHNJAIIH-UHFFFAOYSA-N
SMILESOC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4630 to 1.4670 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid after melting
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link