Phenolphthalein, 98.5%
Phenolphthalein, 98.5%
Thermo Scientific Chemicals

Phenolphthalein, 98.5%

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 147710025
Price (USD)/ Each
482.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
568.00 
Save 85.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
482.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
568.00 
Save 85.35 (15%)
Add to cart
Phenolphthalein, 98.5%
Catalog number147710025
Price (USD)/ Each
482.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
568.00 
Save 85.35 (15%)
-
Add to cart
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only