Thermo Scientific Chemicals

2,4-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 147725000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 147725000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 147725000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-83-2
IUPAC Name
2,4-dichlorophenol
Molecular Formula
C6H4Cl2O
InChI Key
HFZWRUODUSTPEG-UHFFFAOYSA-N
SMILES
OC1=CC=C(Cl)C=C1Cl
Appearance (Color)
White to beige
Appearance (Form)
Crystalline solid
Infrared spectrum
Conforms
Melting point
41°C to 44°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text