2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals
2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 147740100
Price (USD)/ Each
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS513-31-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer0.45 to 0.55 % copper
GC>=75 %
Appearance (Form)Liquid
Appearance (Color)Clear light yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only