Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog number: 147811000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog number: 147811000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 147811000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
407-25-0
IUPAC Name
2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
Molecular Formula
C4F6O3
InChI Key
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)OC(=O)C(F)(F)F
Fluoride (F)
=<50 ppm
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text