Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
250 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 147815000
Price (USD)/ Each
200.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
236.00 
Save 35.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
200.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
236.00 
Save 35.35 (15%)
Add to cart
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Catalog number147815000
Price (USD)/ Each
200.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
236.00 
Save 35.35 (15%)
-
Add to cart
Chemical Identifiers
CAS407-25-0
IUPAC Name2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
Molecular FormulaC4F6O3
InChI KeyQAEDZJGFFMLHHQ-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Sulfate (SO4)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only