Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis

Catalog number: 147850010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis

Catalog number: 147850010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 147850010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6381-92-6
IUPAC Name
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H18N2Na2O10
InChI Key
OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Assay
99.0 to 101.0 % (On dry substance)
pH
4.0 to 6.0 (1 % at 20°C)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text