Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%

CAS: 85715-60-2 | C10H13N2Na3O8 | 358.19 g/mol
Have Questions?
Catalog number 147865000
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.10
Save 18.45 (19%)
Each
-
Add to cart
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.10
Save 18.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS85715-60-2
IUPAC Nametrisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H13N2Na3O8
InChI KeyQZKRHPLGUJDVAR-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Complexometric97.5 to 102.5 % (on dry basis)
Loss on drying=<10 % (1g, 180°C)
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link