3-Amino-9-ethylcarbazole, 90%, tech., Thermo Scientific Chemicals
3-Amino-9-ethylcarbazole, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-9-ethylcarbazole, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147871000
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
Add to cart
3-Amino-9-ethylcarbazole, 90%, tech., Thermo Scientific Chemicals
Catalog number147871000
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS132-32-1
IUPAC Name9-ethyl-9H-carbazol-3-amine
Molecular FormulaC14H14N2
InChI KeyOXEUETBFKVCRNP-UHFFFAOYSA-N
SMILESCCN1C2=C(C=CC=C2)C2=C1C=CC(N)=C2
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Color)Brown
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only