Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)ethane, 98+%, Thermo Scientific Chemicals

Catalog number: 147912500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)ethane, 98+%, Thermo Scientific Chemicals

Catalog number: 147912500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 147912500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1663-45-2
IUPAC Name
[2-(diphenylphosphanyl)ethyl]diphenylphosphane
Molecular Formula
C26H24P2
InChI Key
QFMZQPDHXULLKC-UHFFFAOYSA-N
SMILES
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder or crystals
Melting point
138°C to 144°C
HPLC
>=98.0 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text