2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 147970050
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Molecular FormulaC5H10O4
InChI KeyPTBDIHRZYDMNKB-UHFFFAOYSA-N
SMILESCC(CO)(CO)C(O)=O
Molecular Weight (g/mol)134.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only