p-Toluoyl chloride, 98%, Thermo Scientific Chemicals
p-Toluoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Toluoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 147982500
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.00 
Save 10.35 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.00 
Save 10.35 (15%)
Add to cart
p-Toluoyl chloride, 98%, Thermo Scientific Chemicals
Catalog number147982500
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.00 
Save 10.35 (15%)
-
Add to cart
Chemical Identifiers
CAS874-60-2
IUPAC Name4-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyNQUVCRCCRXRJCK-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration RCOCl>=97.5 %
Refractive index1.5525 to 1.5545 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only