Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 148200250
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
Add to cart
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Catalog number148200250
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
-
Add to cart
Chemical Identifiers
CAS553-53-7
IUPAC Namepyridine-3-carbohydrazide
Molecular FormulaC6H7N3O
InChI KeyKFUSANSHCADHNJ-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
Assay>=96.0 %
Melting point160°C to 163°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only