Sodium formate, 99%
Sodium formate, 99%
Sodium formate, 99%
Thermo Scientific Chemicals

Sodium formate, 99%

CAS: 141-53-7 | CHNaO2 | 68.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 148230010
Price (EUR)
38,85
Online Exclusive
45,70
Save 6,85 (15%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
38,85
Online Exclusive
45,70
Save 6,85 (15%)
Each
Add to cart
Chemical Identifiers
CAS141-53-7
IUPAC Namesodium formate
Molecular FormulaCHNaO2
InChI KeyHLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES[Na+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Melting point253°C to 261°C
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0463019Certificate of AnalysisMay 27, 2024148230051, 148230010
A0457516Certificate of AnalysisNov 20, 2023148230051, 148230010
A0457517Certificate of AnalysisNov 06, 2023148230051, 148230010
A0452974Certificate of AnalysisMay 08, 2023148230051, 148230010
A0451355Certificate of AnalysisMay 02, 2023148230051, 148230010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Label Elements
EU Specific Hazard Statements

May form combustible dust concentrations in air

Share catalog number, name or link

1x1 image pixel for data collection