Thermo Scientific Chemicals

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: 148270025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: 148270025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 148270025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1122-58-3
IUPAC Name
4-(dimethylamino)pyridin-1-ium
Molecular Formula
C7H11N2
InChI Key
VHYFNPMBLIVWCW-UHFFFAOYSA-O
SMILES
CN(C)C1=CC=[NH+]C=C1
Appearance (Form)
Crystalline powder or crystals
Melting point
110°C to 113°C
Water
=<0.1 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text