Thermo Scientific Chemicals

L-5-Hydroxytryptophan, 99%

Catalog number: 148290010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-5-Hydroxytryptophan, 99%

Catalog number: 148290010
1 g, Glass Bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 148290010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4350-09-8
IUPAC Name
(2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Molecular Formula
C11H12N2O3
InChI Key
LDCYZAJDBXYCGN-IMWMWJONNA-N
SMILES
N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
Specific optical rotation
on dry substance
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 % (On dry substance)
Appearance (Color)
White to light beige to light grey
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text