L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
Thermo Scientific Chemicals

L-5-Hydroxytryptophan, 99%

C11H12N2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 148290050
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS4350-09-8
IUPAC Name(2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Molecular FormulaC11H12N2O3
InChI KeyLDCYZAJDBXYCGN-IMWMWJONNA-N
SMILESN[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 % (On dry substance)
Water=<1 % (K.F.)
HPLC=<0.2 % (on dry substance) L-trypthophan
Appearance (Color)White to light beige to light grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link