2-Nitrobenzyl bromide, 97%, Thermo Scientific Chemicals
2-Nitrobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 148310250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3958-60-9
IUPAC Name1-(bromomethyl)-2-nitrobenzene
Molecular FormulaC7H6BrNO2
InChI KeyHXBMIQJOSHZCFX-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
GC>=96.0 %
Appearance (Color)Yellow to light brown
Melting point44°C to 48°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only