α,α,α-Trifluoroacetophenone, 99%, Thermo Scientific Chemicals
α,α,α-Trifluoroacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

α,α,α-Trifluoroacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 148350050
Price (USD)/ Each
43.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.00
Add to cart
α,α,α-Trifluoroacetophenone, 99%, Thermo Scientific Chemicals
Catalog number148350050
Price (USD)/ Each
43.00
-
Add to cart
Chemical Identifiers
CAS434-45-7
IUPAC Name2,2,2-trifluoro-1-phenylethan-1-one
Molecular FormulaC8H5F3O
InChI KeyKZJRKRQSDZGHEC-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4580 to 1.4600 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only