Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Thermo Scientific Chemicals

Benzyl Alcohol, 99%, Pure

CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 L
Packaging:
View all
Metal drum
Catalog number 148390250
Price (USD)
664.65
Special offer
Online exclusive
782.00
Save 117.35 (15%)
Each
-
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
664.65
Special offer
Online exclusive
782.00
Save 117.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Refractive index1.5380 to 1.5410 (20°C, 589 nm)
GC>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470070Certificate of AnalysisMay 17, 2025148390025, 148390250, 148390010
A0465260Certificate of AnalysisAug 14, 2024148390025, 148390250, 148390010
A0452901Certificate of AnalysisMay 03, 2024148390025, 148390250, 148390010
B05040428ACertificate of AnalysisNov 14, 2023148390025, 148390250, 148390010
B0539512ACertificate of AnalysisOct 20, 2023148390025, 148390250, 148390010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection