3-Chloro-2-methylpropene, 90%, tech., Thermo Scientific Chemicals
3-Chloro-2-methylpropene, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-2-methylpropene, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 148420500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS563-47-3
IUPAC Name3-chloro-2-methylprop-1-ene
Molecular FormulaC4H7Cl
InChI KeyOHXAOPZTJOUYKM-UHFFFAOYSA-N
SMILESCC(=C)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4262 to 1.4282 (20°C, 589 nm)
Color scale=<15 APHA
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only