Thermo Scientific Chemicals

Hexyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: 148500025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Hexyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: 148500025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 148500025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-92-7
IUPAC Name
hexyl acetate
Molecular Formula
C8H16O2
InChI Key
AOGQPLXWSUTHQB-UHFFFAOYSA-N
SMILES
CCCCCCOC(C)=O
Appearance (Color)
Clear colorless to light yellow
Specific gravity
(25/25°C) 0.869 to 0.873
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text