Cinnamyl alcohol, 98% trans, Thermo Scientific Chemicals
Cinnamyl alcohol, 98% trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cinnamyl alcohol, 98% trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 148525000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-54-1
IUPAC Name(2Z)-3-phenylprop-2-en-1-ol
Molecular FormulaC9H10O
InChI KeyOOCCDEMITAIZTP-DAXSKMNVSA-N
SMILESOCC=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
GC>=97.5 %
Melting point31°C to 35°C
Appearance (Form)Fused crystalline solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only